1-Amino-cyclobutane-carboxylic acid ethyl ester hydrochloride 96%

Supplier: Ambeed
Warning

A438089-1G A438089-250MG A438089-5G
77016-656EA 67.9 USD
77016-656 77016-658 77016-660
1-Amino-cyclobutane-carboxylic acid ethyl ester hydrochloride 96%
1-Amino-cyclobutane-carboxylic acid ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂·ClH
MDL Number: MFCD05664344
CAS Number: 145143-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR