3-(Trifluoromethyl)-1H-pyrazol-5-amine 97%

Supplier: Ambeed
Warning

A437942-100MG A437942-1G A437942-250MG A437942-25G A437942-5G
77016-646EA 15.92 USD
77016-646 77016-648 77016-650 77016-652 77016-654
3-(Trifluoromethyl)-1H-pyrazol-5-amine 97%
3-(Trifluoromethyl)-1H-pyrazol-5-amine

Precaución: for Research Use Only

Formula: C₄H₄F₃N₃
MW: 151.09 g/mol
MDL Number: MFCD06738292
CAS Number: 852443-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR