1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane 98%

Supplier: Ambeed
Danger

A437640-10G A437640-25G A437640-5G
77004-246EA 6 USD
77004-246 77004-248 77673-070
1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane 98%
1,3,5-Tris[(3,3,3-trifluoropropyl)methyl]cyclotrisiloxane

Precaución: for Research Use Only

Formula: C₁₂H₂₁F₉O₃Si₃
MDL Number: MFCD00054667
CAS Number: 2374-14-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR