Ethyl 3-chloro-4-fluorobenzoate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chloro-4-fluorobenzoic acid ethyl ester
A437122-25G
A437122-5G
76990-580EA
39
USD
76990-580
76990-582
Ethyl 3-chloro-4-fluorobenzoate 95%
Ethyl 3-chloro-4-fluorobenzoate
Precaución: for Research Use Only
Formula:
C₉H₈ClFO₂ |
MDL Number:
MFCD06204341 CAS Number: 137521-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...