(5-Bromo-2-methylphenyl)(5-(4-fluorophenyl)thiophen-2-yl)methanone 95%

Supplier: Ambeed
Warning

A436995-100MG A436995-1G A436995-250MG A436995-5G
77016-630EA 40.32 USD
77016-630 77016-632 77016-634 77016-636
(5-Bromo-2-methylphenyl)(5-(4-fluorophenyl)thiophen-2-yl)methanone 95%
(5-Bromo-2-methylphenyl)(5-(4-fluorophenyl)thiophen-2-yl)methanone

Precaución: for Research Use Only

Formula: C₁₈H₁₂BrFOS
MW: 375.25 g/mol
MDL Number: MFCD25977368
CAS Number: 1132832-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR