(5-Bromo-2-chlorophenyl)boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-chlorophenylboronic acid

A436944-1G A436944-250MG A436944-5G
77004-240EA 199.45 USD
77004-240 77004-242 77004-244
(5-Bromo-2-chlorophenyl)boronic acid 95%
(5-Bromo-2-chlorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrClO₂
MW: 235.27 g/mol
MDL Number: MFCD12026740
CAS Number: 774608-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR