Dithiooxamide 97%

Supplier: Ambeed
Warning

Synonyms: Ethanebis(thioamide), Rubeanic acid

A436451-1G A436451-250MG A436451-25G A436451-5G
77004-232EA 34 USD
77004-232 77004-234 77004-236 77004-238
Dithiooxamide 97%
Dithiooxamide

Precaución: for Research Use Only

Formula: C₂H₄N₂S₂
MDL Number: MFCD00004941
CAS Number: 79-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR