4-Fluoro-1H-indol-6-amine 97%

Supplier: Ambeed
Warning

A435940-100MG A435940-10G A435940-1G A435940-250MG
76990-566EA 119.89 USD
76990-566 76990-568 76990-570 76990-572
4-Fluoro-1H-indol-6-amine 97%
4-Fluoro-1H-indol-6-amine

Precaución: for Research Use Only

Formula: C₈H₇FN₂
MW: 150.153 g/mol
MDL Number: MFCD07357295
CAS Number: 885518-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR