3-Chloro-5-fluorobenzamide 95%

Supplier: Ambeed
Warning

A435740-1G A435740-250MG
76990-556EA 38.2 USD
76990-556 76990-558
3-Chloro-5-fluorobenzamide 95%
3-Chloro-5-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅ClFNO
MW: 173.57 g/mol
MDL Number: MFCD03407964
CAS Number: 874781-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR