(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide 97%

Supplier: Ambeed
Warning

Synonyms: (2-(tert-Butoxy)-2-oxoethyl)triphenylphosphonium bromide

A435674-100G A435674-10G A435674-1G A435674-5G
77033-986EA 45 USD
77033-986 77033-988 77033-990 77033-992
(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide 97%
(tert-Butoxycarbonylmethyl)triphenylphosphonium bromide

Precaución: for Research Use Only

Formula: C₂₄H₂₆BrO₂P
MDL Number: MFCD00075527
CAS Number: 59159-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR