Methyl 2-benzylacrylate 98%

Supplier: Ambeed
Warning

A435395-250MG A435395-5G A435395-100MG A435395-1G
77004-228EA 18 USD
77004-228 77004-230 77673-064 77882-135
Methyl 2-benzylacrylate 98%
Methyl 2-benzylacrylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₂
MDL Number: MFCD07787445
CAS Number: 3070-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR