5-Bromo-3-chloro-2-hydroxybenzaldehyde 95%

Supplier: Ambeed
Danger

A434970-1G A434970-250MG A434970-5G A434970-100MG
76990-548EA 150 USD
76990-548 76990-550 77886-154 77886-153
5-Bromo-3-chloro-2-hydroxybenzaldehyde 95%
5-Bromo-3-chloro-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrClO₂
MDL Number: MFCD00807894
CAS Number: 19652-33-6
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR