Neopentyl glycol diglycidyl Ether 40%
Supplier: Ambeed
|
Warning
|
Synonyms:
Diglycidyloxyneopentane, Neopentyl glycol diglycidyl ether
A434742-100G
A434742-25G
A434742-500G
A434742-1000G
77004-220EA
11
USD
77004-220
77004-222
77004-224
77882-131
Neopentyl glycol diglycidyl Ether 40%
Neopentyl glycol diglycidyl Ether
Precaución: for Research Use Only
Formula:
C₁₁H₂₀O₄ MW: 216.28 g/mol |
MDL Number:
MFCD00040687 CAS Number: 17557-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...