(S)-N-(2,6-Dimethylphenyl)-2-piperidinecarboxamide 98%

Supplier: Ambeed
Warning

Synonyms: (S)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide, N-Despropyl Ropivacaine

A434686-100G A434686-5G
77004-216EA 175.05 USD
77004-216 77004-218
(S)-N-(2,6-Dimethylphenyl)-2-piperidinecarboxamide 98%
(S)-N-(2,6-Dimethylphenyl)-2-piperidinecarboxamide

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O
MW: 232.3214 g/mol
MDL Number: MFCD09038786
CAS Number: 27262-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR