3,5-Diisopropylsalicylic acid 95%

Supplier: Ambeed
Warning

A434576-100G A434576-25G A434576-5G
77016-618EA 219 USD
77016-618 77016-620 77016-622
3,5-Diisopropylsalicylic acid 95%
3,5-Diisopropylsalicylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MDL Number: MFCD00008884
CAS Number: 2215-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR