2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid 95%

Supplier: Ambeed
Warning

Synonyms: BES, 2-(Bis(2-hydroxyethyl)amino)ethanesulphonic acid, N,N-Bis-(2-hydroxyethyl)-2-ethanesulphonic acid

A434319-100G A434319-25G A434319-5G
77033-978EA 23.34 USD
77033-978 77033-980 77033-982
2-[Bis(2-hydroxyethyl)amino]ethanesulfonic acid 95%
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid

Precaución: for Research Use Only

Formula: C₆H₁₅NO₅S
MW: 213.25 g/mol
MDL Number: MFCD00007533
CAS Number: 10191-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR