(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Bis((3aS,8aR)-8,8a-dihydro-3aH-indeno[1,2-d]oxazol-2-yl)methane
A433546-100MG
A433546-1G
A433546-250MG
A433546-25G
A433546-5G
76990-534EA
12.74
USD
76990-534
76990-536
76990-538
76990-540
76990-542
(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole] 98%
(-)-2,2'-Methylenebis[(3aS,8aR)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole]
Precaución: for Research Use Only
Formula:
C₂₁H₁₈N₂O₂ MW: 330.39 g/mol |
MDL Number:
MFCD00799561 CAS Number: 175166-49-1 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...