4-Ethylpiperidine 95%

Supplier: Ambeed
Warning

A433486-100MG A433486-1G A433486-250MG A433486-5G
76990-520EA 63.66 USD
76990-520 76990-522 76990-524 76990-526
4-Ethylpiperidine 95%
4-Ethylpiperidine

Precaución: for Research Use Only

Formula: C₇H₁₅N
MW: 113.20 g/mol
MDL Number: MFCD01872520
CAS Number: 3230-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR