2-Bromo-4-chlorobenzotrifluoride 97%

Supplier: Ambeed
Warning

A433289-1G A433289-5G
77033-968EA 14.86 USD
77033-968 77033-970
2-Bromo-4-chlorobenzotrifluoride 97%
2-Bromo-4-chlorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃
MDL Number: MFCD09839109
CAS Number: 1099597-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR