Ethyl 4-((tert-butyldimethylsilyl)oxy)cyclohexanecarboxylate 95%

Supplier: Ambeed
Warning

A433149-1G A433149-250MG A433149-100MG
76990-516EA 94 USD
76990-516 76990-518 77890-882
Ethyl 4-((tert-butyldimethylsilyl)oxy)cyclohexanecarboxylate 95%
Ethyl 4-((tert-butyldimethylsilyl)oxy)cyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₃₀O₃Si
MDL Number: MFCD24038913
CAS Number: 676560-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR