tert-Butyl (1-benzylazetidin-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A433138-100MG A433138-10G A433138-1G
77033-954EA 6 USD
77033-954 77033-956 77033-958
tert-Butyl (1-benzylazetidin-3-yl)carbamate 95%
tert-Butyl (1-benzylazetidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₂₂N₂O₂
MDL Number: MFCD09878667
CAS Number: 1000577-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR