2-Ethoxy-5-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxypyridine-5-boronic acid pinacol ester, 2-Ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A432830-1G A432830-250MG A432830-5G
77033-946EA 61.54 USD
77033-946 77033-948 77033-950
2-Ethoxy-5-pyridineboronic acid pinacol ester 98%
2-Ethoxy-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₃
MW: 249.12 g/mol
MDL Number: MFCD07781182
CAS Number: 1072945-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR