2-Chloro-4-iodo-6-(trifluoromethoxy)pyridine 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A432709-250MG A432709-1G
77016-604EA 137.92 USD
77016-604 77684-202
2-Chloro-4-iodo-6-(trifluoromethoxy)pyridine 97%, Ambeed.Inc
2-Chloro-4-iodo-6-(trifluoromethoxy)pyridine

Precaución: For research use only.

Formula: C₆H₂ClF₃INO
MW: 323.44 g/mol
MDL Number: MFCD22571776
CAS Number: 1221171-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR