4-Iodo-5-methylpyridin-2-amine 97%

Supplier: Ambeed
Warning

A431394-100MG A431394-1G A431394-250MG
77016-590EA 109.28 USD
77016-590 77016-592 77016-594
4-Iodo-5-methylpyridin-2-amine 97%
4-Iodo-5-methylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₆H₇IN₂
MW: 234.04 g/mol
MDL Number: MFCD16606188
CAS Number: 1227581-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR