1-Allyl-2-bromobenzene 95%

Supplier: Ambeed
Warning

A431381-250MG A431381-5G A431381-1G
76990-504EA 17 USD
76990-504 76990-506 77890-881
1-Allyl-2-bromobenzene 95%
1-Allyl-2-bromobenzene

Precaución: for Research Use Only

Formula: C₉H₉Br
MDL Number: MFCD01319538
CAS Number: 42918-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR