Pyr-βNA 98%

Supplier: Ambeed
Danger

Synonyms: (S)-N-(Naphthalen-2-yl)-5-oxopyrrolidine-2-carboxamide, L-Pyroglutamic acid beta-naphthylamide

A431319-100MG A431319-1G A431319-250MG A431319-5G
77016-582EA 42.44 USD
77016-582 77016-584 77016-586 77016-588
Pyr-βNA 98%
Pyr-βNA

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O₂
MW: 254.28 g/mol
MDL Number: MFCD00055911
CAS Number: 22155-91-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR