(R)-3-Amino-3-(4-chlorophenyl)propanoic acid 97%

Supplier: Ambeed
Warning

A431271-5G A431271-1G A431271-250MG
77004-204EA 321.46 USD
77004-204 77673-056 77673-058
(R)-3-Amino-3-(4-chlorophenyl)propanoic acid 97%
(R)-3-Amino-3-(4-chlorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.63 g/mol
MDL Number: MFCD03840387
CAS Number: 131690-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR