Ethyl 6-(aminomethyl)picolinate hydrochloride 95%

Supplier: Ambeed
Warning

A430860-100MG A430860-250MG A430860-5G
77016-578EA 30.77 USD
77016-578 77016-580 77613-328
Ethyl 6-(aminomethyl)picolinate hydrochloride 95%
Ethyl 6-(aminomethyl)picolinate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃ClN₂O₂
MW: 216.66 g/mol
MDL Number: MFCD19690315
CAS Number: 359015-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR