2-Chloro-4-ethylquinoline-3-carbonitrile 95%

Supplier: Ambeed
Warning

A430325-100MG A430325-1G A430325-250MG
77016-564EA 109.28 USD
77016-564 77016-566 77016-568
2-Chloro-4-ethylquinoline-3-carbonitrile 95%
2-Chloro-4-ethylquinoline-3-carbonitrile

Precaución: for Research Use Only

Formula: C₁₂H₉ClN₂
MW: 216.66 g/mol
MDL Number: MFCD27996494
CAS Number: 101617-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR