1-(2-Amino-4-fluorophenyl)ethanone 95%

Supplier: Ambeed
Warning

A430032-100MG A430032-1G A430032-250MG A430032-5G
77016-556EA 22.28 USD
77016-556 77016-558 77016-560 77016-562
1-(2-Amino-4-fluorophenyl)ethanone 95%
1-(2-Amino-4-fluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈FNO
MW: 153.15 g/mol
MDL Number: MFCD20621031
CAS Number: 159305-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR