n-Octyl-p-toluenesulfonate 97%
Supplier: Ambeed
|
Warning
|
A429903-100G
A429903-1G
A429903-25G
A429903-5G
77033-918EA
139
USD
77033-918
77033-920
77033-922
77033-924
n-Octyl-p-toluenesulfonate 97%
n-Octyl-p-toluenesulfonate
Precaución: for Research Use Only
Formula:
C₁₅H₂₄O₃S |
MDL Number:
MFCD00059447 CAS Number: 3386-35-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...