6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A428930-1G
A428930-5G
77016-540EA
25.47
USD
77016-540
77016-542
6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid 98%
6-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₅ClN₂O₂ MW: 196.59 g/mol |
MDL Number:
MFCD02186559 CAS Number: 182181-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...