(Indolin-4-yl)methanol 97%

Supplier: Ambeed
Warning

A428401-250MG A428401-5G
77033-902EA 100 USD
77033-902 77033-904
(Indolin-4-yl)methanol 97%

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MDL Number: MFCD03095267
CAS Number: 905274-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR