(3-Dimethylaminophenoxy)acetic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(3-(Dimethylamino)phenoxy)acetic acid
A428093-100MG
A428093-1G
A428093-250MG
A428093-50MG
A428093-5G
77016-530EA
11.67
USD
77016-530
77016-532
77016-534
77016-536
77684-193
(3-Dimethylaminophenoxy)acetic acid 98%
(3-Dimethylaminophenoxy)acetic acid
Precaución: For research use only.
Formula:
C₁₀H₁₃NO₃ MW: 195.22 g/mol |
MDL Number:
MFCD06823824 CAS Number: 150188-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...