Ethyl (4-(2-(3-ethyl-4-methyl-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamido)ethyl)phenyl)sulfonylcarbamate 97%

Supplier: Ambeed
Warning

A427966-1G A427966-250MG
77004-194EA 35.01 USD
77004-194 77004-196
Ethyl (4-(2-(3-ethyl-4-methyl-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamido)ethyl)phenyl)sulfonylcarbamate 97%
Ethyl (4-(2-(3-ethyl-4-methyl-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamido)ethyl)phenyl)sulfonylcarbamate

Precaución: for Research Use Only

Formula: C₁₉H₂₅N₃O₆S
MW: 423.48 g/mol
MDL Number: MFCD02955394
CAS Number: 318515-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR