7-Fluoro-1-benzothiophene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 7-Fluorobenzo[b]thiophene-2-carboxylic acid

A427795-1G A427795-5G
77004-190EA 100.79 USD
77004-190 77004-192
7-Fluoro-1-benzothiophene-2-carboxylic acid 97%
7-Fluoro-1-benzothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅FO₂S
MW: 196.2 g/mol
MDL Number: MFCD07377074
CAS Number: 550998-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR