WST-8 95%

Supplier: Ambeed
Danger

Synonyms: 2-(3-(2-Methoxy-4-nitrophenyl)-2-(4-nitrophenyl)-2H-tetrazol-3-ium-5-yl)-5-sulphobenzenesulphonate, sodium salt, 5-(2,4-Disulphophenyl)-3-(2-methoxy-4-nitrophenyl)-2-(4-nitrophenyl)-2H-tetrazolium inner salt monosodium salt

A427585-1MG A427585-250MG A427585-25MG A427585-1G A427585-100MG
77033-896EA 8 USD
77033-896 77033-898 77033-900 77572-326 77872-685
WST-8 95%
WST-8

Precaución: for Research Use Only

Formula: C₂₀H₁₃N₆O₁₁S₂·Na
MW: 600.47 g/mol
MDL Number: MFCD09264687
CAS Number: 193149-74-5
UN: 1325
ADR: 4.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR