2-Fluoro-1-iodo-4-(trifluoromethoxy)benzene 95%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4-iodophenyl trifluoromethyl ether

A427108-100MG A427108-250MG
77016-518EA 35.01 USD
77016-518 77016-520
2-Fluoro-1-iodo-4-(trifluoromethoxy)benzene 95%
2-Fluoro-1-iodo-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃F₄IO
MW: 305.996 g/mol
MDL Number: MFCD20526760
CAS Number: 1321963-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR