(3-Bromo-2-methoxy-5-methylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-methyl-2-methoxyphenylboronic acid

A426321-1G A426321-250MG A426321-5G
77016-512EA 42.44 USD
77016-512 77016-514 77016-516
(3-Bromo-2-methoxy-5-methylphenyl)boronic acid 98%
(3-Bromo-2-methoxy-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BBrO₃
MW: 244.88 g/mol
MDL Number: MFCD04974110
CAS Number: 870717-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR