5-(4-(2-(5-Ethylpyridin-2-yl)ethoxy)benzyl)thiazolidine-2,4-dione hydrochloride 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pioglitazone HCl, 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]-2,4-thiazolidinedione hydrochloride, Pioglitazone hydrochloride
A426266-100G
A426266-1G
A426266-25G
A426266-5G
77004-170EA
206.88
USD
77004-170
77004-172
77004-174
77004-176
5-(4-(2-(5-Ethylpyridin-2-yl)ethoxy)benzyl)thiazolidine-2,4-dione hydrochloride 99%
5-(4-(2-(5-Ethylpyridin-2-yl)ethoxy)benzyl)thiazolidine-2,4-dione hydrochloride
Precaución: for Research Use Only
Formula:
C₁₉H₂₁ClN₂O₃S MW: 392.90 g/mol |
MDL Number:
MFCD04975446 CAS Number: 112529-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...