3-Bromo-4-(piperidin-1-ylmethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A425724-100MG A425724-1G A425724-250MG
76990-476EA 35.01 USD
76990-476 76990-478 76990-480
3-Bromo-4-(piperidin-1-ylmethyl)benzoic acid 95%
3-Bromo-4-(piperidin-1-ylmethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₆BrNO₂
MW: 298.17 g/mol
MDL Number: MFCD11110908
CAS Number: 1131594-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR