3-Amino-4-methyl-N-(4-methyl-2-pyridyl)benzamide 97%

Supplier: Ambeed
Warning

A424804-100MG A424804-1G A424804-250MG
77016-506EA 32 USD
77016-506 77016-508 77016-510
3-Amino-4-methyl-N-(4-methyl-2-pyridyl)benzamide 97%
3-Amino-4-methyl-N-(4-methyl-2-pyridyl)benzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₅N₃O
MDL Number: MFCD11131813
CAS Number: 1019398-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR