(3aR,6aS)-rel-Hexahydro-1H-furo[3,4-c]pyrrole hydrochloride 98%

Supplier: Ambeed
Warning

A424663-100MG A424663-1G A424663-250MG
77016-500EA 83.82 USD
77016-500 77016-502 77016-504
(3aR,6aS)-rel-Hexahydro-1H-furo[3,4-c]pyrrole hydrochloride 98%
(3aR,6aS)-rel-Hexahydro-1H-furo[3,4-c]pyrrole hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO
MW: 149.61 g/mol
MDL Number: MFCD20527059
CAS Number: 57710-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR