5-Bromo-2-phenylthiazole 95%

Supplier: Ambeed
Warning

A423722-100MG A423722-10G A423722-1G
77033-884EA 15.92 USD
77033-884 77033-886 77601-828
5-Bromo-2-phenylthiazole 95%
5-Bromo-2-phenylthiazole

Precaución: for Research Use Only

Formula: C₉H₆BrNS
MW: 240.11 g/mol
MDL Number: MFCD00561163
CAS Number: 53715-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR