(1R,2R)-1-Amino-2-(diphenylphosphino)indane 98%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-2-(Diphenylphosphino)-2,3-dihydro-1H-inden-1-amine

A423419-100MG A423419-1G A423419-250MG
76990-464EA 110 USD
76990-464 76990-466 76990-468
(1R,2R)-1-Amino-2-(diphenylphosphino)indane 98%
(1R,2R)-1-Amino-2-(diphenylphosphino)indane

Precaución: for Research Use Only

Formula: C₂₁H₂₀NP
MDL Number: MFCD17013983
CAS Number: 1091606-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR