L(+)-Glutamic acid hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A422809-1000G
A422809-100G
A422809-10G
A422809-25G
A422809-500G
77004-154EA
34
USD
77004-154
77004-156
77004-158
77004-160
77004-162
L(+)-Glutamic acid hydrochloride 98%
L(+)-Glutamic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₀ClNO₄ |
MDL Number:
MFCD00012619 CAS Number: 138-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...