2-Benzylphenol 97%

Supplier: Ambeed
Warning

A422567-100G A422567-25G A422567-5G
77004-138EA 43 USD
77004-138 77004-140 77004-142 77004-144
2-Benzylphenol 97%
2-Benzylphenol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O
MDL Number: MFCD00002242
CAS Number: 28994-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR