(S)-1-(4-Isopropylphenyl)ethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A422326-100MG A422326-1G A422326-250MG
76990-454EA 108.22 USD
76990-454 76990-456 76990-458
(S)-1-(4-Isopropylphenyl)ethanamine hydrochloride 97%
(S)-1-(4-Isopropylphenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₈ClN
MW: 199.72 g/mol
MDL Number: MFCD16295045
CAS Number: 222737-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR