tert-Butyl-4-(4-nitrobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(4-nitrobenzoyl)piperazine-1-carboxylate

A422308-100MG A422308-1G A422308-250MG A422308-5G
77004-130EA 30.77 USD
77004-130 77004-132 77004-134 77004-136
tert-Butyl-4-(4-nitrobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate 95%
tert-Butyl-4-(4-nitrobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₁N₃O₅
MW: 335.36 g/mol
MDL Number: MFCD06659593
CAS Number: 509073-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR