5-Methyl-3-phenylisoxazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Oxacillin Related Compound C

A422052-1G A422052-5G A422052-25G
77004-114EA 9.55 USD
77004-114 77004-116 77568-848
5-Methyl-3-phenylisoxazole-4-carboxylic acid 98%
5-Methyl-3-phenylisoxazole-4-carboxylic acid

Precaución: For research use only.

Formula: C₁₁H₉NO₃
MW: 203.197 g/mol
MDL Number: MFCD00003153
CAS Number: 1136-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR